Tisk…

Peroxodisíran amonný ≥98%, white crystalline powder for molecular biology

Dodavatel: MP Biomedicals
Nebezpečí

Synonyms: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

0219398825 0219398880 0219398890
ICNA0219398825EA 1550 CZK
ICNA0219398825 ICNA0219398880 ICNA0219398890
Peroxodisíran amonný ≥98%, white crystalline powder for molecular biology
Peroxodisíran amonný

Storage: Store at Room Temperature (15-30 °C), dessicate.
Ammonium persulfate is a strong oxidizing agent. It is a radical initiator.
Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation. APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry. Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, the decolorizing and deodorizing of oils.


  • Molecular Biology Reagent Grade
  • Presentation: White Crystalline Powder
  • Vapor Density: 7.9 (vs air) (lit.)
  • Soluble in water.

Vzorec: (NH₄)₂S₂O₈
Hustota: 950…1050 kg/m³ (bulk density)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS číslo: 7727-54-0
UN: 1444
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR