Tisk…

Cellulose 97.0-102.0%, microcrystalline powder

Dodavatel: MP Biomedicals

0219149980 0219149983 0219149991
ICNA0219149980EA 1130 CZK
ICNA0219149980 ICNA0219149983 ICNA0219149991
Cellulose 97.0-102.0%, microcrystalline powder
Cellulose

Cellulose is an organic compound, a polysaccharide consisting of a linear chain of several hundred to over ten thousand β(1→4) linked D-glucose units.


  • Water soluble substances 12,0 mg/5 g


Cellulose is used as bulking agent, opacifier, anti-caking agent, extrusion aid and stabiliser for foams and emulsions. High purity cellulose powders are used for partition chromatography.


Soluble in ether, insoluble in water (Water soluble substances <12,0 mg/5 g)

Vzorec: (C₁₂H₂₂O₁₁)n
Bod varu: Bulk Density: 0,6 g/ml (25 °C) (Lit,)
Hustota: 0,6 g/ml (25 °C) (lit.)
MDL Number: MFCD00081512
CAS číslo: 9004-34-6

Objednat

Výsledky specifikačního testu

Identity Test Passes Test
Purity 97,0 - 102,0%
Moisture content <5,0%
Particle Size +60 mesh (wt%) <8%
Particle Size +200 mesh (wt%) >45%
pH 5 - 7
Residue on ignition <0,05%
Water Soluble Substances <12,0 mg/5 g
Heavy Metals 10 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR