Tisk…

Amoxicillin

Dodavatel: MP Biomedicals

0219014501 0219014505 0219014510 0219014525
ICNA0219014501EA 1250 CZK
ICNA0219014501 ICNA0219014505 ICNA0219014510 ICNA0219014525
Amoxicillin
Amoxicillin

Amoxycillin is an antibacterial agent against gram-positive and gram-negative bacteria.


Amoxicillin is a moderate-spectrum, β-lactam antibiotic. Amoxicillin is one of the most common antibiotics prescribed for children. It is used to study the oxygen dependent antimicrobial systems of polymorphonuclear leukocytes (PMNLs), the risk of resistance development in Helicobacter pylori, and various dosing strategies against Streptococcus pneumoniae and Pneumococcal pneumonia. It is also used to study the synthesis of bacterial cell walls at the level of peptidoglycan polymer chain cross-linking involving bacterial transpeptidase. Amoxicillin is used to study the synthesis of bacterial cell walls at the level of peptidoglycan polymer chain cross-linking involving bacterial transpeptidase.


Amoxicillin inhibits the synthesis of bacterial cell walls. It inhibits cross-linkage between the linear peptidoglycan polymer chains that make up a major component of the cell walls of both gram-positive and gram-negative bacteria.


Soluble in water (Lit.). Typically used at a concentration of 100 ug/ml and is stable in media at 37 °C for approximately 3 days.

Vzorec: C₁₆H₁₉N₃O₅S
MDL Number: MFCD00056860
CAS číslo: 26787-78-0

Objednat

Výsledky specifikačního testu

Form White Crystalline Powder
Storage Store at +4 °C

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR