Tisk…

4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride), yellow powder

Dodavatel: MP Biomedicals

0215757401 0215757405 0215757410
ICNA0215757401EA 1380 CZK
ICNA0215757401 ICNA0215757405 ICNA0215757410
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride), yellow powder
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

4',6-Diamidino-2-phenylindole Dihydrochloride is a cell permeable fluorescent minor groove-binding probe for DNA.


  • DAPI: excitation: 340 nm; emission: 488 nm (Fluorescence).


It binds to double-stranded DNA by forming a stable complex which fluoresce. DAPI selectively forms complexes with A-T, A-U and I-C base pairs (preferably A-T rich DNA), having one molecule of dye for every 3 base-pairs. DAPI in aqueous solution has two absorbance maxima, at 349 nm and at 263 nm. It can be used as a vital dye to stain nuclei in mature living pollen grains in conjuction with fluroescein diacetate.


Biochem/physiol Actions: Cell permeable fluorescent minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA (preferentially to AT rich DNA), forming a stable complex which fluoresces approximately 20 times greater than DAPI alone.


Slightly soluble in water (10 mg/mL - clear, yellow to yellow-green solution - heat or sonication may be required).

Vzorec: C₁₆H₁₅N₅·2HCl
Storage Temperature: Freezer
MDL Number: MFCD00012681
CAS číslo: 28718-90-3

Objednat

Výsledky specifikačního testu

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR