Tisk…

Probenecid, white powder

Dodavatel: MP Biomedicals
Varování

0215637025 0215637080
ICNA0215637025EA 2850 CZK
ICNA0215637025 ICNA0215637080
Probenecid, white powder
Probenecid

Probenecid is useful as an inhibitor of the organic anion transporter.


Probenecid is an inhibitor of multidrug resistance-associated proteins (Mdr-1). This compound blocks the active site of Mdr-1 and prevents cellular efflux of other active compounds, enhancing their effectiveness by allowing them to remain in the cytoplasm.


Soluble in dilute alkalies; slightly soluble ether and acetone; sparingly soluble in chloroform and ethanol; practically insoluble in water and dilute acids.

Vzorec: C₁₃H₁₉NO₄S
Bod tání: 381…385 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00038402
CAS číslo: 57-66-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR