Tisk…

3-Mercapto-1,2-propanediol ≥98%

Dodavatel: MP Biomedicals
Varování

0215572325
ICNA0215572325EA 1950 CZK
ICNA0215572325
3-Mercapto-1,2-propanediol ≥98%
3-Mercapto-1,2-propanediol

(±)-3-Mercapto-1,2-propanediol (α-Monothioglycerol) is a reagent that is used in analytical chemistry, material science, and cell culture research.

Vzorec: C₃H₈O₂S
Bod varu: 118 °C (1013 hPa)
Hustota: 1,247 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004879
CAS číslo: 96-27-5
EINECS: 202-495-0
Merck Index: 12,09471

Objednat

Výsledky specifikačního testu

Form Clear Liquid
Storage Store at +4 °C, store under nitrogen
Refractive Index n20/D 1,527 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR