Tisk…

Minocycline hydrochloride, powder_yellow

Dodavatel: MP Biomedicals
Varování

Synonyms: (4S,4aS,5aR,12aS)-4,7-Bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride, Minocycline HCl

0215571880 0215571883 0215571891
ICNA0215571880EA 3470 CZK
ICNA0215571880 ICNA0215571883 ICNA0215571891
Minocycline hydrochloride, powder_yellow
Minocycline hydrochloride

Minocycline hydrochloride (INN) is a broad-spectrum tetracycline antibiotic, and has a broader spectrum than the other members of the group. It is a bacteriostatic antibiotic, classified as a long-acting type.


Minocycline inhibits endothelial cell proliferation and angiogenesis.


Minocycline is active against streptococci,enterobacteria, and some mycobacteria, and also against such species as Staphylococcus aureus, Neisseria meningitidis, Acinetobacter, Bacteroides, Haemophilus, and Nocardia.


Minimum inhibitory concentrations (MIC) can range from 0,06 to 1 μg/ml for the most sensitive organisms, and from 4 to 12,5 μg/ml for moderately sensitive organisms.


Soluble in methanol and aqueous solutions of alkali hydroxides and carbonates.

Vzorec: C₂₃H₂₇N₃O₇
MW: 493.94 g/mol
MDL Number: MFCD00083669
CAS číslo: 13614-98-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR