Tisk…

Methyl cellulose, 1500 mPa.S (2% H₂O), fine white powder

Dodavatel: MP Biomedicals

Synonyms: Cellulose methyl ether

0215549580 0215549583 0215549590 0215549591
ICNA0215549580EA 2300 CZK
ICNA0215549580 ICNA0215549583 ICNA0215549590 ICNA0215549591
Methyl cellulose, 1500 mPa.S (2% H₂O), fine white powder
Methyl cellulose

Methyl Cellulose has been used for adhesives or thickening agents, viscosity control agents, or protection in paint formulations. It has also been used as thickeners, binders, emulsifiers, and stabilizers in a variety of cosmetic and food products.


Soluble in cold water. Insoluble in hot water.

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5

Objednat

Výsledky specifikačního testu

Identity Test Passes
Viscosity (2% aq soln, 25°C) 1125 - 2100 cps
Loss on Drying <5.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR