Tisk…

Kyselina 5-aminosalicylová ≥99%, powder

Dodavatel: MP Biomedicals

0215477025 0215477080
ICNA0215477025EA 4690 CZK
ICNA0215477025 ICNA0215477080
Kyselina 5-aminosalicylová ≥99%, powder
Kyselina 5-aminosalicylová

5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .


Solubility: 200 mg plus 4 ml of 1 M HCl. Slightly soluble in cold water and alcohol. More soluble in hot water.

Vzorec: C₇H₇NO₃
MDL Number: MFCD00007877
CAS číslo: 89-57-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR