Tisk…

Ellagic acid, powder

Dodavatel: MP Biomedicals

0215102905 0215102901 0215102910
ICNA0215102905EA 8800 CZK
ICNA0215102905 ICNA0215102901 ICNA0215102910
Ellagic acid, powder
Ellagic acid

Soluble in 1 M Sodium Hydroxide (10 mg/ml - dark brown to green solution) or other alkalies; soluble in pyridine. Slightly soluble in water or ethanol.

Vzorec: C₁₄H₆O₈
MDL Number: MFCD00006914
CAS číslo: 476-66-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR