Tisk…

Eukalyptol ~99%, clear, colourless liquid

Dodavatel: MP Biomedicals
Varování

0215068990 0215068950
ICNA0215068990EA 0 CZK
ICNA0215068990 ICNA0215068950
Eukalyptol ~99%, clear, colourless liquid
Eukalyptol

Eucalyptol (cineole) is an ingredient in many brands of mouthwash and cough suppressant. It is used as an insecticide and insect repellent.It is used in flavorings, fragrances, and cosmetics.


Soluble in alcohol, most fixed oils, organic solvents, glycerin, propylene glycol, chloroform and ether; insoluble in water.

Vzorec: C₁₀H₁₈O
Bod varu: 174…177 °C
Hustota: 0,921…0.924 g/cm³ (20 °C)
Bod vzplanutí: 50 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00167977
CAS číslo: 470-82-6
EINECS: 207-431-5
UN: 1993
ADR: 3,III
Merck Index: 12,03940

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR