Tisk…

L(+)-Norvalin, white crystalline powder

Dodavatel: MP Biomedicals

0210249501 0210249505 0210249510
ICNA0210249501EA 1380 CZK
ICNA0210249501 ICNA0210249505 ICNA0210249510
L(+)-Norvalin, white crystalline powder
L(+)-Norvalin

Enhances NO production from activated macrophages


Soluble in water and 3N-HCl (clear, colorless solution).


Upozornení: Research Use Only (RUO).

Vzorec: C₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS číslo: 6600-40-4

Objednat

Výsledky specifikačního testu

FTIR Conforms to Standard
Optical Rotation (c=2, 6N HCl) +23 ± 5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR