Tisk…

L(+)-Glutamin ≥99%, white crystalline powder

Dodavatel: MP Biomedicals

0210180605 0210180680 0210180683 0210180690 0210180691
ICNA0210180605EA 1030 CZK
ICNA0210180605 ICNA0210180680 ICNA0210180683 ICNA0210180690 ICNA0210180691
L(+)-Glutamin ≥99%, white crystalline powder
L(+)-Glutamin

L-Glutamine, the uncharged and amidated analog of L-glutamic acid, is an important amino acid for the incorporation of NH4+ into biomolecules. It is biosynthesized from NH4+ and glutamate via the enzyme glutamate synthetase. In turn, degradation of glutamine to free the ammonia moiety is mediated by glutaminase. Glutamine also participates in acid-base regulation in vivo.


  • Soluble in water


L-glutamine is an essential amino acid, which serves as a major energy source for cells in culture. L-glutamine is added for optimal cell performance in a culture medium ( Ghyslaine Vanier, (2007), Characterization of the invasion of porcine endothelial cells by Streptococcus suis serotype 2, Canadian Journal Of Camparative Medicine, 71(2), pages: 81-89). The utilization of glutamine under carbon starvation conditions by cultured Saccharomyces cerevisiae and its relation to the cytoplasmic compartmentation of the transcriptional activator Gln3 has been studied. The use of glutamine in cultured rat islets to probe a-ketoisocaproate oxidation has been investigated.


L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.


Soluble in water 41300 mg/l (at 25 °C).


Upozornení: Research Use Only (RUO).

Vzorec: C₅H₁₀N₂O₃
Bod varu: 355 °C (1013 hPa)
Bod tání: 185 °C (lit.)
Hustota: 1,5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS číslo: 56-85-9
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥99%
Residue on Ignition <0.1%
FTIR Conforms to Standard
Moisture <0.5%
Optical Rotation +30 ± 5° (c = 3.5, 1N HCl)
pH (2% aq soln) 4.5-6.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR