Tisk…

4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), tan powder

Dodavatel: MP Biomedicals

Synonyms: 3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid

0210168580 0210168505 0210168525
ICNA0210168580EA 5580 CZK
ICNA0210168580 ICNA0210168505 ICNA0210168525
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), tan powder
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)

Ferulic acid is a chemopreventive caffeic acid analogue. Ferulic acid is an inhibitor of 5-LO (5-lipoxygenase) and 12-LO (12-lipoxygenase).


Ferulic acid is an antioxidant which neutralises free radicals (superoxide, nitric oxide and hydroxyl radical) which could cause oxidative damage of cell membranes and DNA.


Soluble in hot water. (Lit.)

Vzorec: C₁₀H₁₀O₄
MW: 194.18 g/mol
Bod tání: 168…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004400
CAS číslo: 1135-24-6
EINECS: 208-679-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR