Tisk…

Docusate sodium, white waxy solid

Dodavatel: MP Biomedicals
Nebezpečí

Synonyms: Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate

0210157550 0210157590
ICNA0210157550EA 1060 CZK
ICNA0210157550 ICNA0210157590
Docusate sodium, white waxy solid
Docusate sodium

Dioctyl sulphosuccinate sodium salt is used to make a microemulsion with CAPSO for the electrophoresis detection of natural and synthetic estrogens. Used to prepare reverse micelles.


Dioctyl sodium sulphosuccinate (DSS) dissolves slowly in water; at 25 °C to the extent of 1,5 gm/100 cc; at 70 °C, 5,5 gm/100 cc. It dissolves in oils, hydrocarbons, fats and waxs by heating above 75 °C and remains in solution when cooled to room temperature. At room temperature, it is readily soluble in most organic solvents, both polar and non-polar. Soluble in carbon tetrachloride, petroleum ether, naphtha, xylene, dibutyl phthalate, liquid petroleum, acetone, alcohol, vegetable oils.


Upozornení: Research Use Only (RUO).

Vzorec: C₂₀H₃₇NaO₇S
MDL Number: MFCD00012455
CAS číslo: 577-11-7

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 5 - 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR