Tisk…

Cholesterol ≥99%, white powder

Dodavatel: MP Biomedicals

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

0210138201 0210138205 0210138225 0210138280
ICNA0210138201EA 1090 CZK
ICNA0210138201 ICNA0210138205 ICNA0210138225 ICNA0210138280
Cholesterol ≥99%, white powder
Cholesterol

Cholesterol is a component used for the preparation and study of artificial model membranes. One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions. Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential. Cholesterol is suitable for use in liposome preparations. It is standard for clinical work.


Major component of all biological membranes; ~25% of total brain lipid is cholesterol.


Cholesterol monohydrate is practically insoluble in water (less then 0,5 mg/100 ml of water); slightly soluble in alcohol (1,29% w/w at 20 °C), more soluble in hot alcohol (100 g of saturated 96% alcoholic solution contains 28 g at 80 °C). Also one gram dissolves in 2,8 ml of ether, in 4,5 ml of chloroform, and in 1,5 ml of pyridine. The product is soluble in acetone, dioxane, ethyl acetate, benzene, petroleum ether, oils, fats, and in aqueous solutions of bile salts. Solutions should be protected from light.


Upozornení: Research Use Only (RUO).

Vzorec: C₂₇H₄₆O
MW: 386.66
Bod varu: 360
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS číslo: 57-88-5
Merck Index: 13,02221

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥99%
Loss on Drying ≤1.0%
Optical Rotation -34° to -42°

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR