Tisk…

(±)-Aspartic acid ≥99%, white powder

Dodavatel: MP Biomedicals
Varování

0210080383 0210080390 0210080391
ICNA0210080383EA 1350 CZK
ICNA0210080383 ICNA0210080390 ICNA0210080391
(±)-Aspartic acid ≥99%, white powder
(±)-Aspartic acid

DL-Aspartic acid (DL-Asp) is a racemic mixture of the proteinogenic amino acid L-aspartate and the non-proteinogenic amino acid D-aspartate. DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality.


L-Aspartic acid is an endogenous NMDA receptor agonist. Principal neurotransmitter for fast synaptic excitation.


Soluble in NaOH 100 mg/ml.


Upozornení: Research Use Only (RUO).

Vzorec: C₄H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00063083
CAS číslo: 617-45-8

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=1, 1N
HCl)
+21 to +27°
Loss on Drying ≤0.5%
pH 3,0 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR