Tisk…

Kyselina kaprylová ≥99.5%, liquid

Dodavatel: MP Biomedicals
Nebezpečí

0210039191 0210039125 0210039190
ICNA0210039191EA 8920 CZK
ICNA0210039191 ICNA0210039125 ICNA0210039190
Kyselina kaprylová ≥99.5%, liquid
Kyselina kaprylová

Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.
Caprylic acid is an intermediate in the manufacture of perfume esters and of dyes.


It is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil.

Vzorec: C₈H₁₆O₂
Bod varu: 237 °C (lit.)
Bod tání: 15…17 °C (lit.)
Hustota: 0,91 g/cm³ (20 °C) (lit.)
Bod vzplanutí: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS číslo: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity 99.6%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR