Tisk…

Oxid hlinitý, Ecochrom™ Alumina N Activity: Super I

Dodavatel: MP Biomedicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

0204580
ICNA0204580EA 1350 CZK
ICNA0204580
Oxid hlinitý, Ecochrom™ Alumina N Activity: Super I
Oxid hlinitý

Specific Surface Area: 200 m2/g
Particle Size: 63-200 µm
Weight: 0.8 g/ml pH: 7.5


pH: 7.5

Vzorec: Al₂O₃
Hustota: 3,94 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS číslo: 1344-28-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR