Tisk…

N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: BSTFA+TMCS (99:1)

TS-38832
HYPETS-38832EA 4260 CZK
HYPETS-38832
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Vzorec: C₈H₁₈F₃NOSi₂
MDL Number: MFCD00008269
CAS číslo: 25561-30-2
EINECS: 247-103-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR