Tisk…
Aluminium blocks, Reacti-Block™
Dodavatel: Thermo Fisher Scientific
TS-18802 TS-18803 TS-18801 TS-18806 TS-18807 TS-18809 TS-18810 TS-18811 TS-18812 TS-18814 TS-18816 TS-18817 TS-18818 TS-18819
HYPETS-18802EA 5870 CZK
HYPETS-18802 HYPETS-18803 HYPETS-18801 HYPETS-18806 HYPETS-18807 HYPETS-18809 HYPETS-18810 HYPETS-18811 HYPETS-18812 HYPETS-18814 HYPETS-18816 HYPETS-18817 HYPETS-18818 HYPETS-18819
Aluminium blocks, Reacti-Block™
Termostaty blokové Bloky
Reacti-Block™ aluminium blocks are available with many hole configurations, machine-drilled to accommodate almost any size Reacti-Vial™ small reaction vial, test tube or microcentrifuge tube.
  • Made of aluminium alloy for optimal thermal conductivity
  • Block should be fit with sample container for proper heat transfer
Each block contains a thermometer well with size 7,1×36,5 mm (Ø×D mm), except blank blocks J and K.
Aluminium block models A-1, C-1, Z-1, M-1, S-1, U-1 and V-1 can be used with all Reacti-Therm™ modules, including those equipped withMore Product Information