Tisk…
HPLC columns, BetaBasic™
Dodavatel: Thermo Fisher Scientific
71405-104630 71503-154630 71705-254630 71505-254630 71505-154630 71403-154630 71503-153030 71503-104630 71503-054030 71503-034630 71503-014001 71505-014001 71405-014001 71503-052130 71503-012101 71405-052130 71405-154630 71605-154630 71705-052130 71605-052130 71705-014001 71505-052130 71503-102130 71503-054630 71505-104630 71805-254630 71805-154630 71803-012101 71705-154630 71505-014006
HYPE71405-104630EA 28620 CZK
HYPE71405-104630 HYPE71503-154630 HYPE71705-254630 HYPE71505-254630 HYPE71505-154630 HYPE71403-154630 HYPE71503-153030 HYPE71503-104630 HYPE71503-054030 HYPE71503-034630 HYPE71503-014001 HYPE71505-014001 HYPE71405-014001 HYPE71503-052130 HYPE71503-012101 HYPE71405-052130 HYPE71405-154630 HYPE71605-154630 HYPE71705-052130 HYPE71605-052130 HYPE71705-014001 HYPE71505-052130 HYPE71503-102130 HYPE71503-054630 HYPE71505-104630 HYPE71805-254630 HYPE71805-154630 HYPE71803-012101 HYPE71705-154630 HYPE71505-014006
HPLC columns, BetaBasic™
Kolony chromatografické
The BetaBasic™ HPLC columns with its pH stability and with a pore size of 150 Å are ideal for small molecules, peptides and protein digests and are suitable for LC/MS applications.