Tisk…
SPE plates, 96-well, HyperSep™ Diol
Dodavatel: Thermo Fisher Scientific
60300-630 60300-631 60300-632 60300-633
HYPE60300-630EA 13910 CZK
HYPE60300-630 HYPE60300-631 HYPE60300-632 HYPE60300-633
SPE plates, 96-well, HyperSep™ Diol
Mikrodestičky Destičky pro extrakci pevné fáze
The sorbent used in Thermo Scientific™ HyperSep™ Diol Plates features short alkyl chains with polar functional groups, and are ideal for the isolation of polar compounds.
Typical applications include:
- Normal phase extraction of polar compounds
- Purification of polar compounds