Tisk…
Gas Chromatography 3-Port SilFlow Device
Dodavatel: Thermo Fisher Scientific
29063466 29063467 290SF302 60201-390 60201-394 60201-398 60201-401
555-9044EA 2800 CZK
555-9044 555-9043 555-9146 HYPE60201-390 HYPE60201-394 HYPE60201-398 HYPE60201-401
Gas Chromatography 3-Port SilFlow Device
Chromatografické potřeby Chromatography Spare Parts
This SilFlow 3-port device allows column switching in GC-MS without the need to vent.
  • Utilises a make up gas module and a microfluidic device
Informace o dodávce: This device includes a gas module with microfluidics and SilTite™ FingerTite fittings for easy set up and a reliable seal.