Tisk…
Wired Shelves, Norm 5
Dodavatel: HUPFER METALLWERKE
0101852 0101702 0101712 0101822
HUPF0101852EA 11740 CZK
HUPF0101852 HUPF0101702 HUPF0101712 HUPF0101822
Wired Shelves, Norm 5
Nábytek kancelářský Police
Stainless steel wire shelf, electropolished, consisting of 35×10×3,0 mm (1,4×0,4×0,12'') angle sections on the long side. Welded in between are Ø 3,5 mm (0,14'') wires running crosswise at distances of 25 mm (1,0'') with reinforcement struts on the underside made of Ø 3,5 mm ( 0,14'') wires.
  • Well ventilated storage
  • Perfect hygiene
  • High weight capacity
  • Easy handling