Tisk…
Gateway, Profibus
Dodavatel: HUBER
10503
HUBE10503EA 62200 CZK
HUBE10503
Gateway, Profibus
Ovladače Termostaty
This gateway enables the connection of temperature control units to Profibus systems.
Offers a range of possibilities for data communication within SPS and process control systems.
Informace pro objednávání: Includes a mounting bracket and connection cable.