Tisk…
Magnetic stirrer, remote control, Variomag Mobil 10 and 25
Dodavatel: THERMO H + P LABORTECHNIK
50119120
HPLT50119120EA 9870 CZK
HPLT50119120
Magnetic stirrer, remote control, Variomag Mobil 10 and 25
Míchadla Míchačky magnetické
Disc motor drive stirrers and accessory Telemodul controller, which can stir very large liquid volumes and mix highly viscous media. Stirrers have ultra-flat, space-saving design with housing that is easy-to-clean under running water.
  • Remote control unit allows changes to settings to be made easily, even if stirrer is situated in hazardous or inaccessible areas
  • Maintenance-free drive; automatic start-up for safe stirrer acceleration; quick stop function stops stirring in two seconds to prevent vessels breaking
  • Operating conditions for Mobil 10 are −10 to +
More Product Information