Tisk…
Heating tapes, HBRT
Dodavatel: HORST
021502 021500
HORS021502EA 2150 CZK
HORS021502 HORS021500
Heating tapes, HBRT
Ohřívače
These self-limiting heating tape comes with an additional protective braid and humidity proof. It can be used for installations and are cleaned with steam.
  • Nominal temperature 1200 °C
  • With minimum bending radius of 20 mm
  • No starting current required when the heating circuit starts working