Tisk…
Zinc AAS standards, (CRMs), Fluka™
Dodavatel: Honeywell Chemicals
E4085-100ML E4085-500ML E4086-100ML E4086-500ML E4205-100ML E4205-500ML
HONCE4085-100MLEA 958 CZK
HONCE4085-100ML HONCE4085-500ML HONCE4086-100ML HONCE4086-500ML HONCE4205-100ML HONCE4205-500ML
Zinc AAS standards, (CRMs), Fluka™
Standardy Standardy kationické Standardy - zinek
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).