Tisk…
Cerium AAS standards, (CRMs), Fluka™
Dodavatel: Honeywell Chemicals
E4016-100ML E4016-500ML
HONCE4016-100MLEA 1180 CZK
HONCE4016-100ML HONCE4016-500ML
Cerium AAS standards, (CRMs), Fluka™
Standardy Standardy kationické Standardy - cer
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).