Tisk…
Sulphite standard for ion chromatography, (CRM), Fluka™
Dodavatel: Honeywell Chemicals
E3048-100ML
HONCE3048-100MLEA 1970 CZK
HONCE3048-100ML
Sulphite standard for ion chromatography, (CRM), Fluka™
Standardy Standardy anionické Standardy - siřičitan
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).