Tisk…
Benzoate standard for ion chromatography, (CRM), Fluka™
Dodavatel: Honeywell Chemicals
E3004-100ML
HONCE3004-100MLEA 2190 CZK
HONCE3004-100ML
Benzoate standard for ion chromatography, (CRM), Fluka™
Standardy Standardy anionické Standardy - benzoát
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).