Tisk…
Universal Data Acquisition Standard 1 for ICP
Dodavatel: Honeywell Chemicals
E2221-100ML
HONCE2221-100MLEA 8320 CZK
HONCE2221-100ML
Universal Data Acquisition Standard 1 for ICP
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 mg/L each of Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sm, Sc, Tb, Th, Tm, Y, Yb in HNO₃ 5%