Tisk…
NexION KED mode detection limit solution
Dodavatel: Honeywell Chemicals
E2194-100ML
HONCE2194-100MLEA 9310 CZK
HONCE2194-100ML
NexION KED mode detection limit solution
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 g/L each of V, As, Se in HCl 1%