Tisk…
SmartTune solution for standard ELAN/DRC-e Hg for ICP
Dodavatel: Honeywell Chemicals
E2186-100ML
HONCE2186-100MLEA 7610 CZK
HONCE2186-100ML
SmartTune solution for standard ELAN/DRC-e Hg for ICP
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 mg/L each of Ba, Be, Ce, Co, In, Mg, Pb, Rh, U in HNO₃ 1%