Tisk…
Multi-element internal standard for AA and ICP-OES
Dodavatel: Honeywell Chemicals
E2175-100ML
HONCE2175-100MLEA 5790 CZK
HONCE2175-100ML
Multi-element internal standard for AA and ICP-OES
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 mg/L each of Bi, Ho, In, 6Li, Sc, Tb, Y in HNO₃ 2%