Tisk…
Instrument calibration standard 3 for ICP
Dodavatel: Honeywell Chemicals
E2150-100ML
HONCE2150-100MLEA 5440 CZK
HONCE2150-100ML
Instrument calibration standard 3 for ICP
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
1000 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 5%