Tisk…
Instrument calibration standard 2 for ICP
Dodavatel: Honeywell Chemicals
E2149-100ML
HONCE2149-100MLEA 12020 CZK
HONCE2149-100ML
Instrument calibration standard 2 for ICP
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
100 mg/L each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn in HNO₃ 5%