Tisk…
ICP-MS stock tuning solution
Dodavatel: Honeywell Chemicals
E2008-100ML
HONCE2008-100MLEA 5890 CZK
HONCE2008-100ML
ICP-MS stock tuning solution
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 mg/L each of Li, Y, Ce, Co, Tl in HNO₃ 2%