Tisk…
ICP-MS tuning solution
Dodavatel: Honeywell Chemicals
E2007-100ML
HONCE2007-100MLEA 10930 CZK
HONCE2007-100ML
ICP-MS tuning solution
Standardy Standardy kationické Standardy víceprvkové
Certified Reference Material (CRM).
10 mg/L each of Li , Mg, Y, Ce, Tl, Co in HNO₃ 2%