Tisk…
Zinc ICP standards, (CRMs), Fluka™
Dodavatel: Honeywell Chemicals
E1071-100ML E1128-100ML E1198-100ML E1265-100ML
HONCE1071-100MLEA 2220 CZK
HONCE1071-100ML HONCE1128-100ML HONCE1198-100ML HONCE1265-100ML
Zinc ICP standards, (CRMs), Fluka™
Standardy Standardy kationické Standardy - zinek
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).