Tisk…
Palladium ICP standards, (CRMs), Fluka™
Dodavatel: Honeywell Chemicals
E1044-100ML E1115-100ML
HONCE1044-100MLEA 6150 CZK
HONCE1044-100ML HONCE1115-100ML
Palladium ICP standards, (CRMs), Fluka™
Standardy Standardy kationické Standardy - palladium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikace: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).