Tisk…

EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™

Dodavatel: Honeywell Chemicals

33350-100G 33350-6X100G 33350-25KG
33350-100GEA 2230 CZK
33350-100G 33350-6X100G HONC33350-25KG
EDTA magnesium dipotassium salt ≥98% for complexometry, Fluka™
EDTA magnesium dipotassium salt
Vzorec: C₁₀H₁₂K₂MgN₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00054450
CAS číslo: 15708-48-2
EINECS: 239-803-8

Objednat

Výsledky specifikačního testu

Assay Min. 98 %
pH (0.1 M, 20°C) 8.5 - 9.5
Calcium (Ca) Max. 0.05 %
Iron (Fe) Max. 0.001 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR