Tisk…
Tubing for rotarus® peristaltic pumps, Fluran® F-5500-A
Dodavatel: HIRSCHMANN
95220015 95220525 95220522 95220517 95220510 95220017 95220016 95220013
HIRS95220015EA 35240 CZK
HIRS95220015 HIRS95220525 HIRS95220522 HIRS95220517 HIRS95220510 HIRS95220017 HIRS95220016 HIRS95220013
Tubing for rotarus® peristaltic pumps, Fluran® F-5500-A
Hadičky Přenosová hadička
These tubes are for extreme environmental conditions due to its high chemical resistance, resistantance to ozone, sunlight and the weather, extremely tear resistant, high flexibility and elasticity.
  • Temperature resistance from –51 to 204 °C
  • Hardness (Shore durometer A): 60
  • Density: 1,90