Tisk…
Pipette stand, rotary
Dodavatel: HIRSCHMANN
8902101
HIRS8902101EA 2420 CZK
HIRS8902101
Pipette stand, rotary
Stojany na pipety
Pipette stand made from blue PP.
  • Rotatable model with a reinforced base and an additional, rotatable disk to protect the pipette tips
  • 94 holes with different diameters for graduated and volumetric pipettes of any size