Tisk…
Stopcocks for burettes, lateral
Dodavatel: HIRSCHMANN
8901160 8901170 8901175
HIRS8901160EA 1020 CZK
HIRS8901160 HIRS8901170 HIRS8901175
Stopcocks for burettes, lateral
Kohoutky
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
  • Excellent chemical resistance
  • Excellent temperature resistance