Tisk…
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Dodavatel: Avantor
HICHHI-191EA 1940 CZK
HICHHI-191
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Sekáčky
Standard cutter for 1/16" and 1/8" O.D. tubing.
  • Compatible with PEEK and PTFE tubing
  • Flat burr-free cuts
  • Guide holes to ensure precise cutting
Informace o dodávce: Includes five replacement blades.