Tisk…
Size exclusion chromatography columns, InfinityLab AdvanceBio
Dodavatel: Agilent
PL1580-3301 PL1580-1301 PL1180-5350 PL1180-1301 PL1180-5301 PL1180-1350 PL1180-3301 PL1180-3350 PL1580-1350 PL1580-3350 PL1580-5301 PL1580-5350 PL1580-1201 PL1580-3201 PL1580-5201
HEWLPL1580-3301EA 47380 CZK
HEWLPL1580-3301 HEWLPL1580-1301 HEWLPL1180-5350 HEWLPL1180-1301 HEWLPL1180-5301 HEWLPL1180-1350 HEWLPL1180-3301 HEWLPL1180-3350 HEWLPL1580-1350 HEWLPL1580-3350 HEWLPL1580-5301 HEWLPL1580-5350 HEWLPL1580-1201 HEWLPL1580-3201 HEWLPL1580-5201
Size exclusion chromatography columns, InfinityLab AdvanceBio
Kolony chromatografické
These columns are suitable for aggregation analysis via size exclusion chromatography.
  • Increased analytical speed to help you meet vital deadlines
  • Increased resolution for more accurate quantitation
  • Increased sensitivity to quantitate aggregates, even at low levels
  • Increased reproducibility to eliminate rework
AdvanceBio SEC columns deliver accurate, precise quantitation for mAb aggregation and protein SEC analysis.
In addition, AdvanceBio SEC columns improve lab productivity by providing robust, reliable methods that eliminate sample re-analyses.More Product Information