Tisk…
Capillary columns, Porapak Q
Dodavatel: Agilent
G3591-70009 G3591-81066 G3591-81083 G3591-81096 G3591-81135 G3591-81136 G3591-82013 G3591-82016 G3591-82051 G3591-82135 G3591-82136 G3591-81051 G3591-81052 G3591-81053 G3591-81137 G3591-81157 G3591-82053 G3591-82096 G3591-82137 G3591-80013 G3591-80016 G3591-80051 G3591-80052 G3591-80053 G3591-80066 G3591-80083 G3591-80096 G3591-80135 G3591-80136 G3591-80137 G3591-80157 G3591-81013 G3591-81016
HEWLG3591-70009EA 12180 CZK
HEWLG3591-70009 HEWLG3591-81066 HEWLG3591-81083 HEWLG3591-81096 HEWLG3591-81135 HEWLG3591-81136 HEWLG3591-82013 HEWLG3591-82016 HEWLG3591-82051 HEWLG3591-82135 HEWLG3591-82136 HEWLG3591-81051 HEWLG3591-81052 HEWLG3591-81053 HEWLG3591-81137 HEWLG3591-81157 HEWLG3591-82053 HEWLG3591-82096 HEWLG3591-82137 HEWLG3591-80013 HEWLG3591-80016 HEWLG3591-80051 HEWLG3591-80052 HEWLG3591-80053 HEWLG3591-80066 HEWLG3591-80083 HEWLG3591-80096 HEWLG3591-80135 HEWLG3591-80136 HEWLG3591-80137 HEWLG3591-80157 HEWLG3591-81013 HEWLG3591-81016
Capillary columns, Porapak Q
Kolony chromatografické
These Porapak Q columns have wide range of tubing materials including stainless steel, ultimetal and nickel plus hundreds of stationary phases, packings, and supports to suit many specific application.
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance
  • Rigorous, highly efficient packing technology ensures column-to-column reproducibility