Tisk…
HPLC columns, InfinityLab Poroshell 120 HPH-C18
Dodavatel: Agilent
693975-702 693975-502 695975-702 695775-702 695975-502 699975-702 699975-502 699775-702 821725-928 699770-702 823750-930 693770-702 693970-502 693970-702 699970-502 699970-702 699975-502K 699975-702K 693975-502K 693975-702K 695770-702 695775-702K 695970-502 695970-702 695975-502K 695975-702K 699775-702K 690770-702 820750-928 820750-930 821725-930 823750-928 690970-502 690970-702 695675-702
HEWL693975-702EA 32740 CZK
HEWL693975-702 HEWL693975-502 HEWL695975-702 HEWL695775-702 HEWL695975-502 HEWL699975-702 HEWL699975-502 HEWL699775-702 HEWL821725-928 HEWL699770-702 HEWL823750-930 HEWL693770-702 HEWL693970-502 HEWL693970-702 HEWL699970-502 HEWL699970-702 HEWL699975-502K HEWL699975-702K HEWL693975-502K HEWL693975-702K HEWL695770-702 HEWL695775-702K HEWL695970-502 HEWL695970-702 HEWL695975-502K HEWL695975-702K HEWL699775-702K HEWL690770-702 HEWL820750-928 HEWL820750-930 HEWL821725-930 HEWL823750-928 HEWL690970-502 HEWL690970-702 HEWL695675-702
HPLC columns, InfinityLab Poroshell 120 HPH-C18
Kolony chromatografické
Rugged, high-resolution separations up to five times faster than traditional 5 µm methods.
  • High efficiency columns for improved throughput
  • Excellent lot-to-lot reproducibility gives confidence in separation results
InfinityLab Poroshell 120 columns provide exceptional efficiency and reliability, significantly boosting performance from all instruments, whether need improved HPLC performance, UHPLC performance at lower pressures or the highest UHPLC performance. The three particle diameters 1,9 μm, 2,7 μm, and 4 μm allow to choose the size that bestMore Product Information